Learn More
American Research Products Inc Mouse IL13 In Vivo Ab
Supplier: American Research Products Inc IVMB0394100UG
Mouse IL-13 In Vivo Low Endotoxin Antibody [38213] is a in vivo antibody with rat reactivity Available in 100 ug size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles