Learn More
Sino Biological Recombinant Anti-IL-13 R alpha 1/IL13RA1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50088R126100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL13RA1 / IL-13RA1 (rM IL13RA1 / IL-13RA1; Catalog#50088-M08H; NP_598751.3; Met1-Thr340).
Catalog No. 50-255-3470