Learn More
Sino Biological Recombinant Mouse IL-13 R alpha 1/IL13RA1 Protein (ECD, mFc Tag), HPLC-verified
Supplier:  Sino Biological 50088M38H100
Encompass_Preferred
A DNA sequence encoding the mouse IL13RA1 (NP_598751.3) (Met1-Thr340) was expressed with the Fc region of mouse IgG2a at the C-terminus.
Catalog No. 50-255-3473