Learn More
Sino Biological Recombinant Human Carboxypeptidase M Protein (His Tag)
Supplier:  Sino Biological 11228H08H50
Encompass_Preferred
A DNA sequence encoding the human CPM (NP_938079.1) without the propeptide (Met 1-His 422) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6888