Learn More
Sino Biological Recombinant Anti-IL-16 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 51303R005100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL16 (rM IL16; Catalog#51303-M07E; NP_034681.2; Ser1205-Ser1322).
Catalog No. 50-240-7660