Learn More
Sino Biological Recombinant Anti-IL-17A Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 51065R217P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL17 (rM IL17; Catalog#51065-MNAE; Q62386; Thr22-Ala158) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-1272