Learn More
Sino Biological Recombinant Anti-IL-17A Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 51065R21520
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL17 / IL17a (rM IL17 / IL17a; Catalog#51065-MNAE; Q62386; Thr22-Ala158).
Catalog No. 50-241-1263