Learn More
Sino Biological Anti-IL-17A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51065T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL17 (rM IL17; Catalog#51065-MNAE; Q62386; Thr22-Ala158). IL17 specific IgG was purified by Mouse IL17 affinity chromatography.
Catalog No. 50-241-0945