Learn More
Sino Biological Recombinant Mouse IL-17F Protein
Catalog No. 502993333
Encompass_Preferred
A DNA sequence encoding the Mouse IL17F (NP_665855.2) (Arg29-Ala161) was expressed with an initial Met.
Catalog No. 50-299-3333 Supplier Sino Biological Supplier No. 51097MNAE1100
A DNA sequence encoding the Mouse IL17F (NP_665855.2) (Arg29-Ala161) was expressed with an initial Met.