Learn More
Sino Biological Mouse IL17RA MAb 25TS (Sino Biological)
Supplier:  Sino Biological 51065R208P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL17RA (rM IL17RA; Catalog#51065-MNAE; Q62386; Thr22-Ala158) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-237-3787