Learn More
Sino Biological Anti-IL-18BP Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50206T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL18BP / IL18BPa (rM IL18BP / IL18BPa; Catalog#50206-M08H; NP_034661.1; Met1-Ala193). IL18BP / IL18BPa specific IgG was purified by Mouse IL18BP / IL18BPa affinity chromatography.
Catalog No. 50-240-8063