Learn More
Sino Biological Recombinant Anti-IL-18Rα Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50094R032100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL18R1 (rM IL18R1; Catalog#50094-M08H; NP_032391.1; Met1-Gly326).
Catalog No. 50-240-8280