Learn More
Sino Biological Anti-IL-18Rα Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50094RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL18R1 extracellular domain (rM; IL18R1; Catalog#50094-M08H; NP_032391.1; Met 1-Gly 326). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8119