Learn More
Sino Biological Anti-IL-1 alpha/IL1A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50114T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL1A/IL-1A/IL-1F1 (rM IL1A/IL-1A/IL-1F1; Catalog#50114-MNAE; NP_034684.2; Ser115-Ser270). IL1A/IL-1A/IL-1F1 specific IgG was purified by Mouse IL1A/IL-1A/IL-1F1 affinity chromatography.
Catalog No. 50-241-0940