Learn More
Sino Biological Anti-IL-1 beta Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50101T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL1B / IL-1B / IL-1 beta (rM IL1B / IL-1B / IL-1 beta; Catalog#50101-MNAE; P10749; Val118-Ser269). IL1B / IL-1B / IL-1 beta specific IgG was purified by Mouse IL1B / IL-1B / IL-1 beta affinity chromatography.
Catalog No. 50-240-7913