Learn More
Sino Biological Mouse IL1R1 MAb 100TS (Sino Biological)
Supplier:  Sino Biological 50807R191P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-1R1 / CD121a (rM IL-1R1 / CD121a; Catalog#50807-M08H; P13504; Met1-Lys338) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-237-2358