Learn More
Sino Biological Recombinant Anti-IL1R1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50807R191100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-1R1 / CD121a (rM IL-1R1 / CD121a; Catalog#50807-M08H; P13504; Met1-Lys338).
Catalog No. 50-241-1418