Learn More
Sino Biological Mouse IL1R1 Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000786598
Encompass_Preferred
The Mouse IL-1R1 / CD121a ELISA Pair Set is for the quantitative determination of Mouse IL-1R1 / CD121a.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse IL-1R1 / CD121a coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse IL-1R1 / CD121a present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse IL-1R1 / CD121a polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse IL-1R1 / CD121a present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86598 Supplier Sino Biological Supplier No. SEK5080715
The Mouse IL-1R1 / CD121a ELISA Pair Set is for the quantitative determination of Mouse IL-1R1 / CD121a.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse IL-1R1 / CD121a coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse IL-1R1 / CD121a present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse IL-1R1 / CD121a polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse IL-1R1 / CD121a present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.