Learn More
BIOHIPPO Mouse IL1R1 Microsample
SDP
Catalog No. 5000314781
Encompass
Mouse IL1R1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL1R1 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 31 3 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse IL1R1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse IL1R1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse IL1R1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14781 Supplier BIOHIPPO Supplier No. BHE15211985
Mouse IL1R1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL1R1 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 31 3 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse IL1R1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse IL1R1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse IL1R1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color