Learn More
Sino Biological Anti-IL1R1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50807T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL-1R1 / CD121a (Catalog#50807-M08H; P13504; Met1-Lys338). IL-1R1 / CD121a specific IgG was purified by Mouse IL-1R1 / CD121a affinity chromatography.
Catalog No. 50-241-1422