Learn More
Sino Biological Mouse IL1R2 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 51090R016100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-1R2/CD121b (rM IL-1R2/CD121b; Catalog#51090-M08H; P27931; Met1-Glu355).
Catalog No. 50-237-3537