Learn More
Sino Biological Anti-IL1R2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51090T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL-1R2 / CD121b (Catalog#51090-M08H; P27931; Met1-Glu355). IL-1R2 / CD121b specific IgG was purified by Mouse IL-1R2 / CD121b affinity chromatography.
Catalog No. 50-240-8257