Learn More
Sino Biological Mouse IL1RAPL1 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 50900M02H100
Encompass_Preferred
A DNA sequence encoding the mouse IL1RAPL1 (P59823) extracellular domain (Met 1-Thr 357) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-1085