Learn More
Sino Biological Recombinant Anti-ST2/IL-1 RL1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 51143R006100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL1RL1/IL‑1 R4/ST2 (rM IL1RL1/IL‑1 R4/ST2; Catalog#51143-M08H; NP_001020773.1; Met1-Arg332).
Catalog No. 50-241-2089