Learn More
Sino Biological Anti-ST2/IL-1 RL1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51143RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL1RL1 / IL‑1 R4 (rM IL1RL1 / IL‑1 R4; Catalog#51143-M08H; NP_001020773.1; Met1-Arg332). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2088