Learn More
Bon Opus Biosciences Recombinant Mouse Interleukin-2/IL‐2 10ug
SDP
Supplier:  Bon Opus Biosciences CK2410UG
Encompass_Preferred
Recombinant Mouse Interleukin-2 is produced by our E.coli expression system and the target gene encoding Ala21-Gln169 is expressed. Accession# P04351. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM Sodium Citrate, 5% Trehalose, pH4.5. Bon Opus Cat# CK24
Catalog No. 50-217-2818