Learn More
Sino Biological Recombinant Anti-IL-2 Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 51061R449F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Interleukin-2 / IL-2 (rM Interleukin-2 / IL-2; Catalog#51061-MNAE; P04351; Ala21-Gln169) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-241-1423