Learn More
Sino Biological Anti-IL-2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51061RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Interleukin-2 / IL-2 (rM Interleukin-2 / IL-2; Catalog#51061-MNAE; P04351; Ala21-Gln169). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1666