Learn More
Sino Biological Anti-IL-2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51061T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL2/IL-2/Interleukin-2 (rM IL2/IL-2/Interleukin-2; Catalog#51061-MNAE; P04351; Ala21-Gln169). IL2/IL-2/Interleukin-2 specific IgG was purified by Mouse IL2/IL-2/Interleukin-2 affinity chromatography.
Catalog No. 50-241-1207