Learn More
Sino Biological Anti-IL-21 Receptor/IL-21R Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51184RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL-21R (rM IL-21R; Catalog#51184-M08H; AAL82636.1; Met1-Pro236). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2504