Learn More
Proteintech Group Inc Mouse IL22 ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE1004196T
Encompass_Preferred
Interleukin-22 (IL-22) is a recently described IL-10 family cytokine that is produced by T helper (Th) 17 cells, γδ T cells, NKT cells, and newly described innate lymphoid cells (ILCs). IL-22 has the potential to interact with IL-10 because it binds to the IL-10R2c chain with IL-22R1 in its receptor complex. Binding can be blocked by the soluble receptor, IL-22 binding protein (IL-22BP). IL-22 primarily targets nonhematopoietic epithelial and stromal cells, where it can promote proliferation and play a role in tissue regeneration. IL-22 can contribute to immune disease through the stimulation of inflammatory responses, S100s and defensins. IL-22 also promotes hepatocyte survival in the liver and epithelial cells in the lung and gut similar to IL-10.
Catalog No. 50-197-4165