Learn More
Sino Biological Recombinant Mouse IL-23 Protein (His Tag)
Supplier:  Sino Biological CT028M08H50
Encompass_Preferred
A DNA sequence encoding the mouse IL23A (Q9EQ14) (Met1-Ala196 ) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding mouse IL12B (P43432) (Met1-Ser335) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the mouse IL23 heterodimer was purified.
Catalog No. 50-224-7269