Learn More
Sino Biological Recombinant Mouse IL-23 Receptor Protein (hFc Tag)
Catalog No. 502993395
Encompass_Preferred
A DNA sequence encoding the Mouse IL23R (Q8K4B4-1) (Gly24-Gly374,H229Y) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-299-3395 Supplier Sino Biological Supplier No. 51293M02H11
A DNA sequence encoding the Mouse IL23R (Q8K4B4-1) (Gly24-Gly374,H229Y) was expressed with the Fc region of human IgG1 at the C-terminus.