Learn More
Sino Biological Anti-IL-25/IL-17E Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50138T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL25 (rM IL25; Catalog#50138-M07H; NP_542767.1; Val 17-Ala 169). IL25 specific IgG was purified by Mouse IL25 affinity chromatography.
Catalog No. 50-241-2578