Learn More
Sino Biological Recombinant Mouse IL-25/IL-17E Protein (His Tag)
Supplier:  Sino Biological 50138M07H1
Encompass_Preferred
A DNA sequence encoding the mature form of mouse IL25 (NP_542767.1) (Val 17-Ala 169) was expressed with a N-terminal polyhistidine tag.
Catalog No. 50-255-3418