Learn More
Sino Biological Recombinant Mouse IL-27 Protein (His Tag) 1
Supplier:  Sino Biological 51107M08H1
Encompass_Preferred
A DNA sequence encoding the mouse IL27 complex composed of EBI3 (O35228) (Met1-Pro228) and IL27A (Q8K3I6) (Phe29-Ser234) linked by an polypeptide linker was fused with the polyhistidine tag at the C-terminus.
Catalog No. 50-224-6825