Learn More
Sino Biological Mouse IL27Ra Protein 20ug (Sino Biological)
Supplier:  Sino Biological 5A7678M49HB20
Encompass_Preferred
A DNA sequence encoding the Mouse IL27Ra (AAC40121.1) (Met1-Lys510) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-2894