Learn More
Sino Biological Recombinant Anti-CD25/IL-2R alpha Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 50292R051A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD25 / IL2RA (rM CD25 / IL2RA; Catalog#50292-M08H; NP_032393.3; Met1-Lys236) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-240-9105