Learn More
Sino Biological Anti-IL-2R beta/CD122 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50792T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD122/IL2RB (rM CD122/IL2RB; Catalog#50792-M08H; P16297; Met1-Glu240). CD122/IL2RB specific IgG was purified by Mouse CD122/IL2RB affinity chromatography.
Catalog No. 50-241-1663