Learn More
Sino Biological Recombinant Anti-IL-2R gamma/CD132 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50087R011100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD132 / IL2RG (rM CD132 / IL2RG; Catalog#50087-M08H; NP_038591.1; Met1-Ala263).
Catalog No. 50-240-8288