Learn More
Sino Biological Anti-IL-2R gamma/CD132 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50087RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD132 / IL2RG (rM CD132 / IL2RG; Catalog#50087-M08H; NP_038591.1; Met 1-Ala 263). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2144