Learn More
Sino Biological Mouse IL31 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 51070T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL31 (Catalog#51070-M08H; NP_083870.1; Met1-Cys163). IL31 specific IgG was purified by Mouse IL31 affinity chromatography.
Catalog No. 50-237-1621