Learn More
Sino Biological Mouse IL-34 Protein (His Tag) 500ug
Supplier:  Sino Biological 50055M08H500
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Pro 235) of mouse mIL34 (NP_001128572.1) precursor was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6706