Learn More
Sino Biological Anti-IL-36 alpha/IL-1F6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50095RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL-1F6 (Catalog#50095-HNAE; Q9JLA2-1; Met1-His160). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8338