Learn More
Sino Biological Anti-IL-36 beta/IL-1 F8 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51071T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL36B/IL1F8 (rM IL36B/IL1F8; Catalog#51071-MNAE; Q9D6Z6; Met1-Lys183). IL36B/IL1F8 specific IgG was purified by Mouse IL36B/IL1F8 affinity chromatography.
Catalog No. 50-241-1149