Learn More
Sino Biological Anti-IL-36 gamma/IL-1 F9 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51179T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL36G (Catalog#51179-M07E; Q8R460; Gly13-Ser164). IL36G specific IgG was purified by Mouse IL36G affinity chromatography.
Catalog No. 50-240-9637