Learn More
Sino Biological Anti-IL-36 gamma/IL-1 F9 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51179RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL36G (rM IL36G; Catalog#51179-M07E; Q8R460; Gly13-Ser164). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8528