Learn More
Sino Biological Anti-IL-36RN/IL-1F5 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50213RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL-1F5 (Catalog#50213-MNAE; NP_062324.2; Val3-Asp156). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3370