Learn More
Sino Biological Recombinant Mouse IL-4 Protein
Supplier:  Sino Biological 51084MNAE1
Encompass_Preferred
A DNA sequence encoding the mouse IL4 (NP_067258.1) (His23-Ser140) was expressed with a N-terminal Met.
Catalog No. 50-255-3014