Learn More
Sino Biological Anti-IL-4R Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51180T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL-4R / CD124 (Catalog#51180-M08H; NP_001008700.1; Met1-Arg233). IL-4R / CD124 specific IgG was purified by Mouse IL-4R / CD124 affinity chromatography.
Catalog No. 50-240-8529